Learn More
3M™ E-A-RCap™ Headband
Click to view available options
Color: Blue and Yellow; Noise Reduction Rate: 23 db
Description
Protects workers exposed to noise. Ideal disposable hearing protection for factory visitors.
- Round pod plugs that seal the entrance of ear canal, making them more wearer acceptable
- No need to handle the plug tip making them more hygienically acceptable
- Low transmission noise through the band
- Easily stored around the neck when not in use
- Optimum attenuation for many applications
- Easy to wash and clean
- Pods can be easily replaced
Specifications
Specifications
Color | Blue, Yellow |
Certifications/Compliance | CE Approved against European Standard EN352-2:1993. |
Noise Reduction Rating (NRR) | 23 dB |
Plug Shape | Rounded |
Product Type | Ear Plug |
Material | Polycarbonate (Band) / Polyurethane Foam (Ear tip) |