Learn More
E-cadherin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20874-1-AP-20UL
 View more versions of this product
Product Code. 16850455
  • 145.00 € / 20µL
Explore more special offers
Description
Description
This polyclonal antibody recognizes 120 kDa intact E-cadherin and its 80 kDa trypsin-cleaved fragment.
E-Cadherin (epithelial cadherin) is a classical cadherin from the cadherin (alcium dependent adhesion protein) superfamily. The encoded protein is a calcium dependent cell-cell adhesion glycoprotein comprised of five extracellular cadherin repeats, a transmembrane region and a highly conserved cytoplasmic tail. Mutations in this gene are correlated with gastric, breast, colorectal, thyroid and ovarian cancer. Loss of function is thought to contribute to progression in cancer by increasing proliferation, invasion, and/or metastasis. The ectodomain of this protein mediates bacterial adhesion to mammalian cells and the cytoplasmic domain is required for internalization. Identified transcript variants arise from mutation at consensus splice sites. E-cadherin plays a central role in the growth and development of cells by controlling tissue architecture, and maintece of tissue integrity. In humans, E-cadherin is encoded by the CDH1 gene present on chromosome 16. Studies have demonstrated that reduction and/or loss of E-cadherin expression in carcinomas correlates positively with the potential of these tumors for invasion and metastasis.
Specifications
Specifications
E-cadherin
Polyclonal
Unconjugated
CDH1
Arc 1, Cadherin 1, CD324, CDH1, CDHE, E cadherin, ECAD, E-cadherin, LCAM, UVO, Uvomorulin
Rabbit
Antigen Affinity Chromatography
RUO
12550, 83502, 999
-20°C
Liquid
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
0.34 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P09803, P12830, Q9R0T4
Cdh1
E-cadherin Fusion Protein Ag14973
20 μL
Primary
Rat, Human, Mouse