missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences pASG-IBA E. coli Expression Vector with No Secretion
Shop All IBA Lifesciences Products

Klik for at se tilgængelige muligheder
Vector:
pASG-IBA103
pASG-IBA105
pASG-IBA123
pASG-IBA143
pASG-IBA145
pASG-IBA162
pASG-IBA164
pASG-IBA167
pASG-IBA168
pASG-IBA23
pASG-IBA3
pASG-IBA43
pASG-IBA45
pASG-IBA5
pASG-IBA62
pASG-IBA64
pASG-IBAwt1
Pakningsstørrelse:
5µg
Beskrivelse
- The pASG-IBA vectors are designed for expression of target proteins without affinity tag in E. coli.
- The vector carries an ampicillin resistance cassette for selection of transformed E. coli cells, F1 and ColE1 origin for a high plasmid copy number, and the inducible tetracycline promoter/operator for the regulated expression of proteins.
- The vectors can be combined with any E. coli strain since the tet-promoter works independently of the genetic background of E. coli.
- Expression of the recombinant protein is induced by addition of anhydrotetracycline, which is a derivative of tetracycline that does not exhibit antibiotic activity.
Tekniske data
Tekniske data
| Inducing Agent | Inducible with anhydrotetracycline |
| Marker Gene | Ampicillin |
| Promoter | Tet |
| Product Type | Vector |
| Content And Storage | Recommended: 2°C to 8°C for frequent usage, -20°C for long-term storage |
| Species | E.coli |
| Protein Tag | Twin-Strep-tag, GST Tag |
| Cloning Method | Direct Cloning |
| Concentration | 250 ng/μL |
| Primer | Forward |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion