Learn More
Invitrogen™ E-Gel™ EX Double Comb Agarose Gels
Invitrogen E-Gel EX Double Comb precast agarose gels are designed for ultrasensitive and convenient DNA sample electrophoresis in as little as 5 to 8 minutes.
258.00 € - 950.00 €
Specifications
Gel Type | E-Gel |
---|---|
Product Type | Agarose Gel |
Stain Type | SYBR™ Gold |
Wells | 22-well |
Well Design | Double Comb |
Product Code | Brand | Gel Percentage | Separation Range | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Gel Percentage | Separation Range | Quantity | Price | Quantity & Availability | |||
17115632
|
Invitrogen™
A42345 |
1% | 100 to 5000 bp | 10 Gels |
259.00 €
Pack of 10 |
In Stock
Log in to see stock. |
|||
17126378
|
Invitrogen™
A44887 |
1% | 100 to 5000 bp | 2 x 10 Gels |
421.00 €
Pack of 20 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||
17136378
|
Invitrogen™
A44888 |
1% | 100 to 5000 bp | 5 x 10 Gels |
950.00 €
Pack of 50 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||
16865321
|
Invitrogen™
A42346 |
2% | 50 to 2000 bp | 10 Gels |
258.00 €
Pack of 10 |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||
16875321
|
Invitrogen™
A44889 |
2% | 50 to 2000 bp | 2 x 10 Gels |
418.00 €
Pack of 20 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||
16885321
|
Invitrogen™
A44890 |
2% | 50 to 2000 bp | 5 x 10 Gels |
948.00 €
Pack of 50 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||
Description
Invitrogen E-Gel EX Double Comb precast agarose gels are designed for ultrasensitive and convenient DNA sample electrophoresis in as little as 5–8 minutes. Each E-Gel EX Double Comb gel cassette contains all components and stain required for efficient gel separation and analysis—just load your samples and run. E-Gel EX Double Comb gels are ideal for analyzing PCR products, restriction digests, plasmid preparations, and genotyping samples.
Features of E-Gel EX Double Comb Agarose Gels include:
- Instantaneous setup—precast E-Gel agarose gels are ready for use when you are, and do not require gel preparation or additional buffers
- High sensitivity—detect 1 ng of sample DNA
- Rapid analysis—complete DNA sample separation in 5–8 minutes
- In-process control—view DNA sample migration in real time
- Easy sample access—openable cassette design allows easy in-gel access for convenient DNA sample excision or transfer to membranes
- For DNA—ideal for DNA sample analysis
Ultra-sensitive detection of DNA
E-Gel EX Double Comb gels include SYBR Gold DNA stain and were developed for ultimate sensitivity, demonstrating up to five-fold greater sensitivity than comparable gels containing ethidium bromide. This superior sensitivity allows you to use lower amounts of sample, saving time and money.
Fast and convenient analysis
E-Gel EX Double Comb gels are our fastest-resolving agarose gels, enabling sample analysis in as little as 5-8 minutes. Each E-Gel EX Double Comb gel contains agarose, electrodes, SYBR Gold DNA stain, and a bufferless ion exchange system packaged inside a dry disposable cassette. E-Gel technology does not required any gel or buffer preparation or gel staining steps. Just load your samples and run.
Live monitoring of DNA migration
E-Gel EX Double Comb gels include SYBR Gold II DNA gel stain with an excitation wavelength in the blue-light spectrum. When used on the E-Gel Power Snap Electrophoresis Device with integrated blue-light trans-illuminator, less-hazardous real-time monitoring of DNA migration is enabled without risk to eyes or possible damage of the sample, unlike UV illumination.
Easy access to the gel
E-Gel EX Double Comb cassettes are designed for convenient opening with a Gel Knife (Cat. No. EI9010), for easy excision of specific bands or transfer of the gel to a membrane for southern blot analysis.
Running and imaging devices
E-Gel EX Double Comb gels require the Invitrogen E-Gel Power Snap Electrophoresis System for gel running and viewing. The system consists of an electrophoresis device and a camera for fast and convenient E-Gel EX Double Comb gel separation and analysis.
Available in a variety of gel formats
E-Gel agarose gels are available in variety of gel percentage, stain, and well formats. The 22-well E-Gel EX Double Comb gels are available in 1% and 2% gel percentages.
Specifications
E-Gel | |
SYBR™ Gold | |
Double Comb |
Agarose Gel | |
22-well |
For Research Use Only. Not for use in diagnostic procedures.