Learn More
Invitrogen™ E-Gel™ Power Snap Electrophoresis Device Starter Kit, CloneWell II 0.8%
Designed for fast and convenient E-Gel™ agarose gel electrophoresis, from DNA sample loading to gel image. Includes E-Gel™ CloneWell™ II agarose gel, 0.8%.
Brand: Invitrogen™ G8168ST
1330.73 EUR valid until 2025-03-21
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Includes: E-Gel Power Snap Electrophoresis device,orange viewing glasses,E-Gel Go! agarose gel adaptor,region specific cable with power adaptor.
Description
The Invitrogen E-Gel Power Snap Electrophoresis Device Starter Kit with 0.8% E-Gel CloneWell II agarose gels provides everything you need for faster and more effective DNA fragment isolation in a cloning workflow.
The kit includes:
- 10 E-Gel CloneWell II agarose gels with SYBR Safe, 0.8% (Cat. No. G661818)
- E-Gel Power Snap Electrophoresis Device (Cat. No. G8100)
- E-Gel 1 Kb Plus Express DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 10488091)
Services
1 year warranty

Specifications
• 1 electrophoresis device • 10 E-Gel CloneWell II gels • 1 vial E-Gel 1 Kb Plus Express DNA Ladder • 1 power adapter • 1 pair of viewing glasses • 1 E-Gel Go! Adapter Store at room temperature. |
|
1 kit | |
48 VDC |
Electrophoresis System Starter Kit | |
Room Temperature | |
240 VAC |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.