missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ Power Snap Electrophoresis Device Starter Kit, DC Safe 1%

The Invitrogen™ E-Gel™ Power Snap Electrophoresis Device Starter Kit, DC Safe 1%, provides everything you need to quickly start using precast E-Gel™ Double Comb Agarose Gels with SYBR Safe DNA Gel Stain on the device.

Brand:  Invitrogen™ G8171ST

1387.34 EUR valid until 2024-12-15
Use promo code "21658" to get your promotional price.


Product Code. 17185979

  • 1954.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The kit includes:

  • Ten E-Gel™ Double Comb Agarose Gels with SYBR Safe DNA Gel Stain, 1% (Cat. No. A42347)
  • E-Gel™ Power Snap Electrophoresis Device (Cat. No. G8100)
  • E-Gel™ 1-Kb Plus Express DNA Ladder with E-Gel™ Sample Loading Buffer (1X) (Cat. No. 10488091)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

• Ten E-Gel Double Comb Agarose Gels with SYBR Safe DNA Gel Stain, 1% (Cat. No. A42347)
• E-Gel Power Snap Electrophoresis Device (Cat. No. G8100)
• E-Gel 1-Kb Plus Express DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 10488091)

Store at room temperature.
E-Base™, E-Gel™ Imager
Electrophoresis System Starter Kit
1 kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.