missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ Power Snap Electrophoresis Device Starter Kit, DC Safe 2%

The Invitrogen™ E-Gel™ Power Snap Electrophoresis Device Starter Kit, DC Safe 2%, provides everything you need to quickly start using precast E-Gel™ Double Comb Agarose Gels with SYBR Safe DNA Gel Stain on the device.

Brand:  Invitrogen™ G8172ST

1380.24 EUR valid until 2024-12-15
Use promo code "21658" to get your promotional price.


Product Code. 17195979

  • 1944.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The kit includes:

  • Ten E-Gel™ Double Comb Agarose Gels with SYBR Safe DNA Gel Stain, 2% (Cat. No. A42348)
  • E-Gel™ Power Snap Electrophoresis Device (Cat. No. G8100)
  • E-Gel™ Low Range Quantitative DNA Ladder with E-Gel™ Sample Loading Buffer (1X) (Cat. No. 12373031)
     
TRUSTED_SUSTAINABILITY
Specifications

Specifications

• Ten E-Gel Double Comb Agarose Gels with SYBR Safe DNA Gel Stain, 2% (Cat. No. A42348)
• E-Gel Power Snap Electrophoresis Device (Cat. No. G8100)
• E-Gel Low Range Quantitative DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 12373031)

Store at room temperature.
E-Base™, E-Gel™ Imager
Electrophoresis System Starter Kit
1 kit
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.