missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ Power Snap Electrophoresis Device Starter Kit, SYBR Safe 2%

Provides everything needed to quickly start using precast 2% E-Gel agarose gels with SYBR Safe on the device.

Brand:  Invitrogen™ G8122ST

1110.20 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.


Product Code. 17750469

  • 1820.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Provides everything needed to quickly start using precast 2% E-Gel agarose gels with SYBR Safe on the device

Includes

  • 10 E-Gel agarose gels with SYBR Safe, 2% (A42135)
  • E-Gel Power Snap Electrophoresis Device (G8100)
  • E-Gel 50 bp DNA Ladder with E-Gel Sample Loading Buffer, 1X (10488099)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

• 1 electrophoresis device
• 10 E-Gel agarose gels with SYBR Safe, 2%
• 1 vial E-Gel 50 bp DNA Ladder
• 1 power adapter
• 1 pair of viewing glasses
• 1 E-Gel Go! Adapter

Store at room temperature.

Electrophoresis Device Starter Kit
Room Temperature
Mini
1 kit
240 V AC (Power Supply), 48 V DC (Electrophoresis Device)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.