missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ E-Gel™ Power Snap Electrophoresis Device Starter Kit, SizeSelect II 2%
Designed for fast and convenient E-Gel™ agarose gel electrophoresis, from DNA sample loading to gel image. Includes E-Gel™ SizeSelect™ II agarose gel, 2%.
Brand: Invitrogen™ G8162ST
1373.14 EUR valid until 2024-12-15
Use promo code "21658" to get your promotional price.
Includes: E-Gel Power Snap Electrophoresis device,orange viewing glasses,E-Gel Go! agarose gel adaptor,region specific cable with power adaptor.
Description
The Invitrogen E-Gel Power Snap Electrophoresis Device Starter Kit with 2% E-Gel SizeSelect II agarose gels provides everything you need for faster and more effective DNA fragment library size selection in a next-generation sequencing (NGS) workflow.
The kit includes:
- 10 E-Gel SizeSelect II agarose gels, 2% (Cat. No. G661012)
- E-Gel Power Snap Electrophoresis Device (Cat. No. G8100)
- E-Gel Sizing DNA Ladder (Cat. No. 10488100)
Services
1 year warranty
Specifications
• 1 electrophoresis device • 10 E-Gel SizeSelect II gels • 1 vial E-Gel Sizing DNA Ladder • 1 power adapter • 1 pair of viewing glasses • 1 E-Gel Go! Adapter Store at room temperature. |
|
1 kit | |
240 V AC (Power Supply), 48 V DC (Electrophoresis Device) |
Electrophoresis Device Starter Kit | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.