Learn More
Invitrogen™ E-Gel™ Power Snap Electrophoresis System Starter Kit, EX 1%
System starter for fast and convenient E-Gel™ agarose gel electrophoresis, from DNA sample loading to gel image. Includes E-Gel EX 1% agarose gels.
Brand: Invitrogen™ G8341ST
Additional Details : Weight : 1.10000kg
Includes: E-Gel Power Snap Electrophoresis device,E-Gel Power Snap Camera,orange viewing glasses,E-Gel Go! agarose gel adaptor,region specific cable with power adaptor.
Description
The Invitrogen E-Gel Power Snap Electrophoresis System Starter Kit, EX 1%, provides everything you need to quickly start using precast E-Gel EX 1% agarose gels on the system for a streamlined gel electrophoresis and gel imaging workflow.
The kit includes:
- 10 E-Gel EX agarose gels, 1% (Cat. No. G401001)
- E-Gel Power Snap Electrophoresis System (Cat. No. G8300)
- E-Gel 1 Kb Plus Express DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 10488091)
- Gel opening knife (Cat. No. EI9010)
Services
1 year warranty
Specifications
• 1 electrophoresis device • 1 camera device • 10 E-Gel EX 1% gels • 1 vial E-Gel 1 Kb Plus Express DNA Ladder • 1 power adapter • 1 pair of viewing glasses • 1 E-Gel Go! Adapter • 1 knife Store at room temperature. |
|
Electrophoresis System Starter Kit | |
240 V AC (Power Supply), 48 V DC (Electrophoresis Device) |
Mini | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.