missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ Power Snap Electrophoresis System Starter Kit, SYBR Safe 2%

Provides everything needed to quickly start using precast 2% E-Gel agarose gels with SYBR Safe on the system for a streamlined gel electrophoresis and gel imaging workflow.

Brand:  Invitrogen™ G8322ST

2453.05 EUR valid until 2025-03-28
Use promo code "24107" to get your promotional price.


Product Code. 17856531

  • 3455.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Provides everything needed to quickly start using precast 2% E-Gel agarose gels with SYBR Safe on the system
  • For a streamlined gel electrophoresis and gel imaging workflow

Includes

  • 10 E-Gel agarose gels with SYBR Safe, 2% (A42135)
  • E-Gel Power Snap Electrophoresis System (G8300)
  • E-Gel 50 bp DNA Ladder with E-Gel Sample Loading Buffer, 1X (10488099)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

• 1 electrophoresis device
• 1 camera device
• 10 E-Gel agarose gels with SYBR Safe, 2%
• 1 vial E-Gel 50 bp DNA Ladder
• 1 power adapter
• 1 pair of viewing glasses
• 1 E-Gel Go! Adapter

Store at room temperature.

1 kit
48 VDC
Electrophoresis System Starter Kit
Room Temperature
240 VAC
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.