missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E-Gel™ Power Snap Electrophoresis System Starter Kit, SYBR Safe 1%

Provides everything needed to quickly start using precast 1.2% E-Gel agarose gels with SYBR Safe on the system for a streamlined gel electrophoresis and gel imaging workflow.

Brand:  Invitrogen™ G8321ST

2314.60 EUR valid until 2024-12-15
Use promo code "21658" to get your promotional price.


Product Code. 17751478

  • 3260.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Provides everything needed to quickly start using precast 1.2% E-Gel agarose gels with SYBR Safe on the system
  • For a streamlined gel electrophoresis and gel imaging workflow

Includes

  • 10 E-Gel agarose gels with SYBR Safe, 1% (A42100)
  • E-Gel Power Snap Electrophoresis System (G8300)
  • E-Gel 1 Kb Plus DNA Ladder with E-Gel Sample Loading Buffer, 1X (10488090)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

• 1 electrophoresis device
• 1 camera device
• 10 E-Gel agarose gels with SYBR Safe, 1%
• 1 vial E-Gel 1 kb Plus DNA Ladder
• 1 power adapter
• 1 pair of viewing glasses
• 1 E-Gel Go! Adapter

Store at room temperature.

1 kit
240 V AC (Power Supply), 48 V DC (Electrophoresis Device)
Electrophoresis System Starter Kit
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.