missing translation for 'onlineSavingsMsg'
Learn More

Heidolph Instruments e-Lift

Comfortable and secure work with the electric lab jack e-Lift.

Brand:  Heidolph Instruments 509-00100-00

Product Code. 17956692

  • 1655.00 € / Each
Estimated Shipment: 16-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Thanks to the adjusted plate (190 mm x 400 mm), the magnetic stirrer does not wobble or slip.
  • A winch for manual operation and an attachment for additional support rods on the lab jack are both included - for a secure fixing of your setups.
  • The height can be adjusted from 100 mm to 275 mm which corresponds to a vertical lift of 175 mm.
  • Compatible with the Hei-PLATE Mix 'n' Heat Ultimate magnetic stirrer.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Metal
Jack Lift
Hei-PLATE Mix 'n' Heat Ultimate Magnetic Stirrer
100 to 275 mm
Autoclavable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Heidolph Instruments e-Lift > Material: Metal

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.