Learn More
Invitrogen™ E-PAGE™ Midi Protein Gels, 3.7 mm
E-PAGE™ Gels are self-contained, pre-cast gels that include a gel matrix and electrodes packaged inside a disposable cassette.
582.00 € - 706.00 €
Specifications
For Use With (Equipment) | E-BASE |
---|---|
Gel Size | Midi |
Gel Thickness | 3.7 mm |
Gel Type | E-PAGE |
Separation Type | Denaturing |
Product Code | Brand | Gel Percentage | Wells | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Gel Percentage | Wells | Price | Quantity & Availability | ||||
10654583
|
Invitrogen™
EP09606 |
6% | 96-well |
706.00 €
Each |
Estimated Shipment: 06-02-2025
Log in to see stock. |
||||
10206172
|
Invitrogen™
EP04808 |
8% | 48-well |
582.00 €
Each |
Estimated Shipment: 18-04-2025
Log in to see stock. |
||||
Beschreibung
E-PAGE Gels are self-contained, pre-cast gels that include a gel matrix and electrodes packaged inside a disposable cassette. Protein separation on an E-PAGE Gel requires an EBase Integrated Device, a compact and automated platform designed for medium- and high-throughput electrophoresis.
Each E-PAGE 8% 48-well gel contains 48 sample lanes and 4 marker lanes with a 3.2-cm separation length. E-PAGE 48-well gels can be loaded with a 12- or 8-channel pipettor or 4- or 8-tip robotic system (Figure 1).
Each E-PAGE 6% 96-well gel contains 96 sample lanes and 8 marker lanes with a 1.6-cm separation length. The well openings of the cassette are compatible with the standard 96-well plate format and can be conveniently loaded with a multichannel pipettor or 8-, 12-, or 96-tip liquid-handling robotic device (Figure 2).
Both E-PAGE 48 and 96 cassettes can be opened to remove the gel for downstream staining or blotting (Figures 3 and 4). In addition, each cassette is labeled with an individual barcode to facilitate identification of the gel using commercial barcode readers.
More about the E-PAGE precast gel system
The Invitrogen E-PAGE precast gel system is designed for fast, mess-free medium- and high-throughput protein electrophoresis. It consists of the E-PAGE precast gels, E-Base Integrated devices for running the gels, and free E-Editor 2.0 software for image analysis.
The all-in-one E-Gel Power Snap Plus Electrophoresis System is designed for fast and convenient high-throughput gel electrophoresis. With dry precast E-PAGE gel technology, you can run protein samples in as little as 14 minutes. The device arrives with pre-programmed protocols for both E-PAGE gel types.
Spezifikation
E-BASE | |
3.7 mm | |
Denaturing | |
E-PAGE™ | |
Room Temperature | |
8 cm |
Midi | |
E-PAGE | |
Molecular Weight | |
8 Gels/Box | |
Store at room temperature, do not allow the temperature to drop below 4°C or rise above 40°C | |
13 cm |
Sicherheit und Handhabung
missing translation for 'shelfLife' : 6 Months