Learn More
Thermo Scientific™ E1-ClipTip™ Bluetooth™ Electronic Adjustable Tip Spacing Multichannel Equalizer Pipettes
Perform sample transfers between virtually any tube, rack, microplate or horizontal gel box quickly and efficiently with adjustable tip spacing.
Brand: Thermo Scientific™ 4672030BT
Description
Perform sample transfers between virtually any tube, rack, microplate or horizontal gel box quickly and efficiently with the Thermo Scientific™ E1-ClipTip™ Equalizer Pipette. Adjustable tip spacing allows you to set the distance between tips by simply sliding the scale to expand or contract to the desired labware format.The NEW My Pipette™ Creator app enables programming of E1-ClipTip from the comfort of your PC.
The following products are recommended for SARS Cov-2 testing: 4672100BT and 4672060BT.
Innovative ClipTip interlocking tip interface technology*- Tip is locked and sealed in place until ejected — no dropped tips that waste valuable samples
- Complete seal to secure the sample volume in each tip for enhanced accuracy and precision
- Tips are attached and ejected with a light touch — eliminates the frustration of banging tips on the pipette
My Pipette Creator to revolutionize the way you pipette with a new era of technology
- New web-based pipetting app powered by Thermo Fisher Cloud
- Enables efficient, centralized programming and sharing of protocols between E1-ClipTip pipettes and colleagues
- Offers pre-programmed protocols, to download and share, for many of your favorite reagent kits and pipetting protocols
- Connect E1-ClipTip pipette to the app via a Bluetooth** or USB connection.
- Accelerate collaboration and reproducibility, all from the comfort of your computer.
E1-ClipTip pipette to help you enjoy pipetting
- E1-ClipTip is designed for optimum comfort and ease of use: the electronic tip ejection and index finger pipetting action let your thumb relax during pipetting.
- Two operating options: Step-based Matrix or Presets with settings for your most common applications.
- The Programs function allows you to store 30 your most common Matrix or Presets protocols with a specific name.
- Connect your E1-ClipTip pipette to My Pipette Creator app conveniently via Bluetooth**
- Five different user profiles offer customized settings for multiple users and protocols
- MyPipette™ function to customize your main menu view by organizing the icons in your preferred order or creating shortcuts for your favorite programs and pipetting functions. You can also protect important settings from unintentional editing with a password.
- Service and calibration trackers remind you when the pipette needs to be sent for service
- Li-ion battery for long lasting performance and fast recharging. Recharging with a charging stand or a plug, you decide.
- 2-year warranty with convenient web registration
E1-ClipTip™ Equalizer: transferring multiple samples between different labware formats
- Adjustable tip spacing allows you to set the distance between pipette tips for different types of labware simply by sliding a scale to expand or contract the tip spacing to the desired format:
- Microcentrifuge tubes to/from 24,-48,-96-and 384- well microplates
- Test tubes to/from deepwell blocks
- Microplates and tubes to agarose gels
- Save up to 90% in time and repetitions compared to use of single channel manual pipettes***
E1-ClipTip pipette and ClipTip tips for every application
- Choose your E1-ClipTip, volume range from 0.5 μL to 1250μL:
- Single channels
- Multichannels with 8, 12 or 16 channels in 96 and 384 formats
- Adjustable tip spacing multichannels with 6, 8, 12 channels in 96 and 384 formats
- Wide selection of ClipTip tips:
- ClipTip 96-format tips are available in 20, 50, 200, 300, 1000 and 1250 μl
- ClipTip 384-format tips are available in 12.5, 30 and 125 μl
- Several packaging options: filtered, sterile, non-sterile, racked and reload
* E1-ClipTip 12.5 and all 384 E1-ClipTip pipettes utilize snap and seal technology.
** Bluetooth models available in following countries/regions: Andorra, Australia, Austria, Belgium, Bulgaria, Canada, China, Croatia, Cyprus, Czech Republic, Denmark, Estonia, Finland, France, Germany, Greece, Hong Kong, Hungary, Iceland, India, Ireland, Italy, Japan, Korea (Rep. of), Latvia, Liechtenstein, Lithuania, Luxembourg, Malta, Monaco, Netherlands, New Zealand, Norway, Poland, Portugal, Romania, San Marino, Singapore, Slovakia, Slovenia, Spain, Sweden, Switzerland, Taiwan, United Kingdom, United States and Vatican. For other countries catalog numbers without 'BT' extension should be used.
*** Compared to a manual single channel pipette filling a 96-well microplate from microcentrifuge tubes. See Thermo Scientific Smart Note: Is there an easier and more efficient way to transfer liquids between various labware formats, than using traditional handheld pipettes? 2014.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified. The Bluetooth™ is a trademark of Bluetooth SIG, Inc.
Specifications
ClipTip 384 30 | |
Violet | |
Bluetooth | |
E1-ClipTip™ | |
Equalizer Pipette | |
0.01μL |
4.5 to 14.2mm | |
ClipTip™ 384 30 | |
8 | |
Yes | |
1 to 30 μL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.