Learn More
Thermo Scientific™ E1-ClipTip™ Electronic Pipette Charging Stand
Conveniently charge and store one or three pipettes with these custom charging stands.
Brand: Thermo Scientific™ 9420510
549.48 EUR valid until 2024-09-27
Use promo code "21716" to get your promotional price.
This item is not returnable.
View return policy
Description
Conveniently charge and store one or three E1-ClipTip pipettes with the Thermo Scientific™ E1-ClipTip™ Charging Stands.
Warranty: 1 yearSpecifications
E1-ClipTip Charging Stand, for 3-pipettes | |
E1-Clip Tip Electronic Single and Multichannel Pipette | |
E1-ClipTip Electronic single and multichannel pipettes | |
Includes universal power supply |