missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EAPP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16931-1-AP-150UL
This item is not returnable.
View return policy
Description
This protein may play an important role in the fine-tuning of both major E2F1 activities, the regulation of the cell-cycle and the induction of apoptosis. Promotes S-phase entry, and inhibits p14(ARP) expression. The E2F family plays a crucial role in the control of cell cycle and action of tumor suppressor proteins and is also a target of the transforming proteins of small DNA tumor viruses. The E2F proteins contain several evolutionally conserved domains found in most members of the family. These domains include a DNA binding domain, a dimerization domain which determines interaction with the differentiation regulated transcription factor proteins (DP), a transactivation domain enriched in acidic amino acids, and a tumor suppressor protein association domain which is embedded within the transactivation domain. The C-terminal of EAPP binds the N-terminal of E2F1 and also interacts with E2F2 and E2F3, but not E2F4.Specifications
EAPP | |
Polyclonal | |
Unconjugated | |
EAPP | |
BM036, C14orf11, E2F associated phosphoprotein, EAPP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
299043, 55837, 66266 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q56P03, Q5BU09 | |
EAPP | |
EAPP Fusion Protein Ag10403 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |