Learn More
Hoefer™ Easy Breeze™ Gel Dryer Accessories: Frame Components and Accessories
For use with the SE 1200 Easy Breeze Gel Drying System
75.86 €
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
11505604
|
Hoefer™
SE1213 |
Black Quarter-turn Knobs for Standard or Large Frame Assemblies | N/A | ||||||
Description
- Highly resistant to acids, bases, and all commonly used buffer systems
- Easily removed while maintaining gel integrity
- Lightweight and virtually unbreakable when used with reasonable care
Specifications
SE 1200 Easy Breeze Gel Drying System |