missing translation for 'onlineSavingsMsg'
Learn More

H-B Instrument™ Easy-Read™ General Purpose Liquid-In-Glass Thermometers: Partial Immersion

Easy-to-read partial immersion thermometers

27.58 € - 32.30 €

Specifications

Certifications/Compliance Traceable to NIST. Includes a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.
For Use With (Application) Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Material Glass
Thermometer Fill Enviro-Safe™ Liquid
View More Specs

Products 3
Product Code Brand Subdivision Length (Metric) Overall Temperature Range (English) Temperature Range (Metric) Price Quantity & Availability  
Product Code Brand Subdivision Length (Metric) Overall Temperature Range (English) Temperature Range (Metric) Price Quantity & Availability  
16419623
View Documents
H-B Instrument™
B603010300
1°C 300 mm n.a. -20°C to +150°C
32.30 €
Each
Estimated Shipment: 24-04-2025
to see stock.
Add to basket
 
16401265
View Documents
H-B Instrument™
B60302-0000
1°C 350 mm n.a. -10°C to +260°C
31.84 €
Each
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
 
15265053
View Documents
H-B Instrument™
B60301-0100
0.5°C 300 mm n.a. -35°C to +50°C
27.58 €
Each
Estimated Shipment: 28-04-2025
to see stock.
Add to basket
 
Description

Description

  • Lead-free, yellow back glass and black liquid provides easy readability and fewer reading errors
  • Contains non-toxic, biodegradable, Enviro-Safe™ liquid
  • EnviroKleen™ certified

Applications:

Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.

Order Info

Should you require an individual Thermometer Calibration Report, factory calibrated thermometers or custom calibration services are available.

Certifications

Manufactured and tested by our USA quality team, these thermometers are traceable to NIST™ and include a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.

Specifications

Specifications

Traceable to NIST. Includes a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy and Traceability.
Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Glass
Enviro-Safe™ Liquid
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.