Learn More
Thermo Scientific™ EASY-Spray™ Nano & Capillary Emitters
Use this easy-to-use sprayer in your sensitive LC-MS separation of protein and peptides using nano and capillary LC.
Brand: Thermo Scientific™ ES791A
Description
Connect your nano or capillary column to your Thermo Scientific™ mass spectrometers (MS) more easily using Thermo Scientific™ EASY-Spray™ Nano & Capillary Emitters. The nano and capillary emitters act as a column-independent sprayer, allowing the introduction of low flow from nano and capillary columns without troublesome handling of traditional spray needles. There are two versions of emitters available: with and without integrated transfer lines. These sprayers are ideal for protein and peptide separation using nano and capillary HPLC.
For sensitive protein and peptide separation using nano and capillary HPLC
These emitters offer easy connection to Thermo Scientific™ MAbPac™ and Accucore™ nano and capillary columns for enhanced separation and MS detection of proteins, peptides and mnoclonal antibodies (mAbs).
Specifications
EASY-Spray Emitter | |
Transfer line: 20 μm I.D. x 50 cm Length | |
Easy Spray | |
Emitter |
0.02 mm | |
500 mm | |
Unpacked | |
Nano EASY-Spray Emitter with transfer line |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.