missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Easy-Titer™ Mouse IgG Assay Kit

Determine antibody concentration in serum and culture supernatant with kits specific for mouse, rabbit, or human IgG, and human IgM.

Brand:  Thermo Scientific™ 23300

484.50 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.


Product Code. 10454365

  • 518.00 € / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Easy-Titer Mouse IgG Assay Kit
Solution-based Detection, Absorbance
96 Tests
Colorimetric
Target-specific
Sufficient For: 96 total tests (including 9 standards per use)
• Goat Anti-Mouse IgG Sensitized Beads, 2mL
• Easy-Titer Dilution Buffer, 30 mL
• Easy-Titer Blocking Buffer, 15 mL

Store at 4°C (do not freeze).
Spectrophotometer, Microplate Reader
Protein Quantitation Assay, Isotyping Assay, Immunoassay
Easy-Titer™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZI399886 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number23300
Lot NumberZF393944 Certificate TypeCertificate of Analysis Date04/07/2024 Catalog Number23300
Lot NumberZA371214 Certificate TypeCertificate of Analysis Date30/01/2024 Catalog Number23300
Lot NumberYJ376164 Certificate TypeCertificate of Analysis Date04/11/2023 Catalog Number23300
Lot NumberYA367770 Certificate TypeCertificate of Analysis Date04/02/2023 Catalog Number23300
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Easy-Titer™ Mouse IgG Assay Kit > Easy-Titer™ Mouse IgG Assay Kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.