Learn More
3M™ Easy Trap Duster Sheets
Trap and hold eight times more dust, dirt, sand and hair than traditional mops and other disposable dusting systems. Two sided disposable sheets, works in wet environments and on most horizontal surfaces. Low drag reduces worker fatigue.
Brand:  3M™ 7100081546
 View more versions of this product
Product Code. 17287790
  • 208.00 € / 1 roll
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Duster system traps and holds six times more dust, dirt, sand and hair than traditional mops
  • Low drag reduces worker fatigue
  • Works in wet environments
  • Works on most horizontal surfaces
  • Two sided disposable sheets for use with your current dusting tools
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Duster Sheets
203 x 152 mm
60 Sheets/Roll, 8 Rolls/Cs.