Learn More
BRAND™ EASYCAL™ 5.0 Server License
Server license of the EASYCAL 5.0 calibration software with free updates.
Brand: BRAND™ 708414
This item is not returnable.
View return policy
Description
The EASYCAL™ 5 calibration software facilitates the monitoring of Liquid Handling instruments (like piston-stroke devices such as pipettes, dispensers, burettes, and manual dispensers), as well as volumetric instruments made of glass or plastic, based on GLP/GMP and DIN EN ISO 9001. EASYCAL™ 5 software can be used for BRAND instruments as well as instruments from other manufacturers.
- Automatically calculates and compares all values against the tolerances from current standards or individually-defined limits (tolerances of numerous devices and the interface settings of over 100 test devices (e.g. balances) are already stored for you)
- Choose between a stand-alone license for working on one workstation or a client/server version for parallel, distributed work on multiple workstations
- Stand-alone license is recommended for small labs where calibration is completed by one person
- Client/server version and its floating licenses are ideal for larger labs where calibration will be carried out at multiple workstations
- For testing of liquid handling devices and volumetric instruments made of glass and plastic according to standards such as ISO 8655, ISO 4787, and others
- Open software, suitable for all volumetric instruments - from any manufacturer
- Extensive library with device specifications from many well-known manufacturers can be expanded and modified by the user to help you get started and minimize data entry time
- Quickly and easily search and find the owner, test history, and next test date
- Continuous control of current actual state during testing by graphical representations and real time calculation of statistical values
- Reminder function for upcoming tests with automatic email notifications sent to device owner
- Integration of address data of your customers and suppliers in a business partner database
- User management with user roles (e.g., inspector, supervisor, and system administrator) and access restriction to EASYCAL functions
- Dual-control principle for the release of critical data such as test plans, calibration orders before certificate printing, and device specification
- RS-232 interface connection for measuring devices such as scales, thermometers, barometers, and hygrometers with automatic data transmission of the measured values
- Customize certificates and audit trails to your needs in the integrated certificate editor
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Calibration Software | |
Calibration for Large Laboratories |