Learn More
STEMCELL Technologies EasySep™ Mouse NK Cell Isolation Kit
Immunomagnetic isolation of untouched mouse NK cells
Brand:  STEMCELL Technologies 19855
Encompass
Product Code. 17197971
  • 924.00 € / 1 set
Estimated Shipment: 27-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The EasySep™ Mouse NK Cell Isolation Kit targets non-NK cells by labeling unwanted cells with biotinylated antibodies and EasySep™ Streptavidin RapidSpheres™, and separates cells without columns using an EasySep™ magnet
  • Desired cells are simply poured off into a new tube
  • Isolated cells are immediately available for downstream applications such as flow cytometry, culture or cell-based assays
  • This product replaces the EasySep™ Mouse NK Cell Enrichment Kit (Catalog #19755) for even faster cell isolations
Specifications
Specifications
NK Cells
Cell Isolation
1 x 109 cells