Learn More
STEMCELL Technologies EasySep™ Mouse T Cell Isolation Kit
Immunomagnetic isolation of untouched mouse T cells
Brand:  STEMCELL Technologies 19851
Encompass
Product Code. 16617665
  • 886.50 € / Each
Estimated Shipment: 27-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The EasySep™ Mouse T Cell Isolation Kit is designed to isolate T cells from single-cell suspensions of splenocytes or other tissues by negative selection
  • Unwanted cells are targeted for removal with biotinylated antibodies directed against non-T cells and streptavidin-coated magnetic particles
  • Labeled cells are separated using an EasySep™ magnet without the use of columns
  • Desired cells are poured off into a new tube
  • This product replaces the EasySep™ Mouse T Cell Enrichment Kit (Catalog #19751) for even faster cell isolations
Specifications
Specifications
T Cells
Cell Isolation
1 x 109 cells