Learn More
STEMCELL Technologies EasySep™ Rat B Cell Isolation Kit
Immunomagnetic isolation of untouched rat B cells
Brand:  STEMCELL Technologies 19644
Encompass
Product Code. 17107941
  • 975.00 € / 1 set
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The EasySep™ Rat B Cell Isolation Kit is designed to isolate B cells from single-cell suspensions of splenocytes or lymph nodes or from whole blood by negative selection
  • The EasySep™ procedure involves labeling unwanted cells with antibody complexes and magnetic particles
  • The magnetically labeled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube
Specifications
Specifications
B Cells
Cell Isolation
1 x 109 cells