missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EasyStrip™ Plus Tube Strip with Attached Ultra Clear Caps

The Thermo Scientific™ EasyStrip™ Plus Tube Strip with Attached Ultra Clear Caps is designed to reduce the risk of cross-contamination in qPCR assays.

Brand:  Thermo Scientific™ AB2005

195.96 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.


Product Code. 15606154

  • 426.00 € / Pack of 250

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: 250 strips
This item is not returnable. View return policy

Description

Description

The Thermo Scientific™ EasyStrip™ Plus Tube Strip with Attached Ultra Clear Caps is designed to reduce the risk of cross-contamination in qPCR assays. The tube strip is compatible with 0.2mL thermal cycler blocks and features individually attached optical caps for quick and easy access to individual samples. In addition, graduated 20μL markers on each tube enable visual volume verification and measurement of reagent and sample additions.

For tube strips with flat caps, please see Cat. No. AB-2000.

Features:
  • Avoid cross contamination with the individually attached caps
  • Verify tube reaction volumes with graduated 20μL markings on each tube
  • Track samples easily using the end tabs to label each strip
  • Identify individual wells quickly and prevent pipetting errors using the labels (A-H) on each tube cap and cap hinge (visible when cap is open and closed)
Applications
  • qPCR/Real-Time PCR

For Research Use Only. Not for use in diagnostic procedures.

Certifications

Certified human DNA, RNase and DNase free

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Certified DNA/RNase Free
0.2 mL
250 strips
EasyStrip™ 
Room Temperature
250 Strip Tubes w/ Attached Caps
Natural, Ultra Clear
0.25 mL
Thermal Cyclers or Real-Time PCR Systems
Polypropylene
Ultra Clear Flat Caps
8-Strip Tube
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers