missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Essential Human T-Cell Phenotyping Kit
Click to view available options
Quantity:
100 reactions
Unit Size:
100 reactions
Description
Phenotyping Kit leverages high-quality eBioscience™ antibodies and isotype controls and includes a ready-to-use protocol for experimental setup, gating strategy, and analysis.
Benefits include:
- Proven performance―antibodies and supporting reagents designed for flow cytometry
- Highly referenced antibodies―eBioscience™ portfolio contains trusted fluorochrome-conjugated antibodies and reagents for immunology and oncology research
- Turnkey kit and protocol―contains reagents and standardized gating recommendations needed to perform flow cytometry characterization
Specifications
Specifications
| Content And Storage |
|
| Format | Kit |
| Quantity | 100 reactions |
| Detection Method | Fluorescence |
| Shipping Condition | Approved for shipment at Room Temperature or on Wet or Dry Ice |
| Product Line | eBioscience |
| Product Type | Phenotyping Kit |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction