Learn More
Invitrogen™ eBioscience™ Mouse Cell Surface Staining Flow Cytometry Workflow Kit
The eBioscience™ Mouse Cell Surface Flow Cytometry Workflow Kit contains all of the normal buffers, blockers, and beads needed to complete a standard flow cytometry experiment for staining cell surface targets on mouse cell suspensions.
Brand:  Invitrogen™ A53012
Product Code. 17787463
  • 333.00 € / Each
Explore more special offers
Description
Description
  • Kit contains all of the normal buffers, blockers, and beads to complete a standard flow cytometry experiment for staining cell surface targets on mouse cell suspensions
  • When combined with experiment-specific antibodies, the reagents within this kit have been optimized to work together to detect mouse cell surface targets by flow cytometry
  • Effective compensation controls
  • Fc receptor blocking
  • Non-antibody, non-protein based blocking solution for reducing background staining from fluorophores
  • Antibody binding buffer which minimizes non-specific binding of antibodies
  • Fixation buffer for storage of stained samples
Kit Includes
  • UltraComp eBeads™ Plus Compensation Beads (01-3333-41)
  • CD16/CD32 Monoclonal Antibody (93), eBioscience (14-0161-81)
  • CellBlox™ Blocking Buffer (B001T06F01)
  • eBioscience™ Flow Cytometry Staining Buffer (00-4222-57)
  • eBioscience™ IC Fixation Buffer (00-8222-49)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 Kit
Flow Cytometry