missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Ebony Treatment Dish
Protects DNA from damage. Fisherbrand™ Ebony Treatment Dishes are manufactured with a darkened acrylic to reduce exposure to light that may be harmful to DNA.
94.05 € - 296.00 €
Specifications
Color | Ebony |
---|---|
For Use With (Equipment) | Comet Assay Tank |
Material | Acrylic |
Product Code | Brand | No. per Pack | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. per Pack | Price | Quantity & Availability | |||||
15589901
|
Fisherbrand™
STAINDISH |
1 Each |
94.05 €
Each |
Estimated Shipment: 03-02-2025
Log in to see stock. |
|||||
15599901
|
Fisherbrand™
STAINDISH4X |
4 |
296.00 €
Pack of 4 |
Estimated Shipment: 03-02-2025
Log in to see stock. |
|||||
Description
- Manufactured from Ebony Acrylic
- Minimizes exposure to light and reduces background DNA damage
Specifications
Ebony | |
Comet Assay Tank | |
Acrylic |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title