Learn More
Ebrotidine, MedChemExpress
MedChemExpress Ebrotidine(FI 3542) is a competitive H2-receptor antagonist (Ki= 127.5 nM) with a potent antisecretory activity and evidenced gastroprotection.
Brand: Medchem Express HY-15538
Description
Ebrotidine(FI 3542) is a competitive H2-receptor antagonist (Ki= 127.5 nM) with a potent antisecretory activity and evidenced gastroprotection.Specifications
Ebrotidine | |
Off-White | |
C14H17BrN6O2S3 | |
FI3542 | |
O=S(C1=CC=C(Br)C=C1)(/N=C/NCCSCC2=CSC(NC(N)=N)=N2)=O | |
477.42 | |
Research | |
COVID-19-immunoregulation |
100981-43-9 | |
Research | |
5 mg | |
DMSO : 100 mg/mL (209.46 mM; Need ultrasonic) | |
477.42 | |
99.43% | |
Solid |