missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ EBSS, calcium, magnesium, phenol red
GREENER_CHOICE

Earle's Balanced Salt Solution (EBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents.

Brand:  Gibco™ 24010043

Product Code. 11540616

  • 25.57 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Earle's Balanced Salt Solution (EBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation.

This EBSS is modified as follows:

With: Glucose, Sodium Bicarbonate, Calcium, Magnesium, Phenol Red

The complete formulation is available.

Product Intended Use

For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: 15-30° C.
Shipping conditions: Ambient.
Shelf life: 36 months from date of manufacture.
1 X
Earle's Balanced Salt Solution (EBSS)
Phenol Red, Glucose, Sodium Bicarbonate, Calcium, Magnesium
cGMP-compliant under the ISO 13485 standard
7 to 7.4
500 mL
Animal Origin-free
Liquid
Sterile-filtered
No Sodium Pyruvate
270 - 310 mOsm/kg
Gibco™
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.