Learn More
Trajan Scientific and Medical™ EC Science Silicon Septa
Designed for long lifetime and low injection port adhesion.
Brand:  Trajan Scientific and Medical™ 041905
 View more versions of this product
Product Code. 15238623
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Combines long injection lifetime, low bleed and low injection port adhesion
  • Reliable seal against carrier gas pressure in the inlet
  • No bleed or contamination into the GC system
  • Maximum operating temperature of 350°C (300°C for 17mm size)
Specifications
Specifications
EC
50/Pk.