missing translation for 'onlineSavingsMsg'
Learn More

ECL Plex Goat anti-Mouse IgG-Cy™ 3 Secondary Antibody, Cytiva

Brand:  Life Science products Cytiva PA43010

 View more versions of this product

Product Code. 11515075

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Host Species: Goat
Isotype: IgG
Species Reactivity: Ms

Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

IgG
Cyanine3
600 μg
Purified
Monoclonal
Goat
Secondary
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.