missing translation for 'onlineSavingsMsg'
Learn More

ECL Plex Goat anti-Rabbit IgG-Cy™ 3 Secondary Antibody, Cytiva

222.09 € - 483.00 €

Specifications

Antigen IgG
Classification Monoclonal
Conjugate Cyanine3
Form Purified
Host Species Goat
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11580634
View Documents
Life Science products Cytiva
28-9011-06
150 μg
483.00 €
150µg
Please to purchase this item. Need a web account? Register with us today!
11590634
View Documents
Life Science products Cytiva
28-9011-07
600 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Host Species: Goat
Isotype: IgG
Species Reactivity: Rb

Western Blotting

Specifications

Specifications

IgG
Cyanine3
Goat
Secondary
Monoclonal
Purified
IgG
Videos
Safety and Handling

Safety and Handling

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P273-Avoid release to the environment.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
ECL Plex Goat anti-Rabbit IgG-Cy™ 3 Secondary Antibody, Cytiva

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.