missing translation for 'onlineSavingsMsg'
Learn More

ECL Plex Goat anti-Rabbit IgG-Cy™ 5 Secondary Antibody, Cytiva

166.57 € - 190.00 €

Specifications

Antigen IgG
Classification Monoclonal
Conjugate Cyanine5
Form Purified
Host Species Goat
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11545075
View Documents
Life Science products Cytiva
PA45011
150 μg
190.00 €
150µg
Please to purchase this item. Need a web account? Register with us today!
11555075
View Documents
Life Science products Cytiva
PA45012
600 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Host Species: Goat
Isotype: IgG
Species Reactivity: Rb

Western Blotting

Specifications

Specifications

IgG
Cyanine5
Goat
Secondary
Monoclonal
Purified
IgG
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3

Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H317-May cause an allergic skin reaction.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.