missing translation for 'onlineSavingsMsg'
Learn More

Labcon™ Eclipse™ 200 μL Wide Orifice Pipette Tips

These beveled point tips have a 1.45 mm orifice for use with viscous samples.

Brand:  Labcon™ 1026-960-006-9

 View more versions of this product

Product Code. 17567786

  • 59.40 € / Pack of 960
Estimated Shipment: 19-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Fits most standard 200 μL pipettes
  • Polypropylene USP Class VI Resin
  • Made in USA
Specifications

Specifications

200 μL
Non-sterile
Purple
1.95 in.
ISO 9001:2015, FDA CFR21 GMP, USP Class VI
Liquid Handling
Wide Orifice, Bevel Point
Single-use
No
Polypropylene Resin
Non-filtered
Clear
Autoclavable
49.7 mm
ATP, RNase/DNase, Endotoxin-free
Rack, 96
960 Tips/Pk.
Universal Fit
Eclipse
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers