Learn More
Sharpsguard™ Eco 1 Waste Container
GREENER_CHOICE
The new Sharpsguard™ eco 1 container, made using at least 40% recycled plastic that is used for the disposal of sharps that have been contaminated with medicines, but not cytotoxic.
Brand:  Sharpsguard™ DD477OLR
 View more versions of this product
Product Code. 17774854
  • 14.92 € / Each
Edge
Explore more special offers
Description
Description
  • Sharpsguard™ eco 1 is ideal for phlebotomists. Its push-down, one-piece lid incorporates a vacutainer remover, and will fit securely into our POUDS™ tray range and other carrying trays.
  • This container is already supplied with the lid assembled. Details about how to lock the container and keep the users and contents safe can be found on the label.
  • This container is for disposal by incineration or other authorized process and is designed for the consignment of waste categorized by EWC codes: 18-01-01, 18-01-03, 18-02-01, 18-02-02.
Specifications
Specifications
0.84 L
204 x 68 x 119 mm
154 g
Orange
1 Ea.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Sharpsguard™ Eco 1 Waste Container > Waste Container; Color: Orange