Learn More
Sharpsguard™ Eco 11.5 Waste Container
GREENER_CHOICE
The new Sharpsguard™ eco containers, made using at least 40% recycled plastic that is used for the disposal of sharps that have not been contaminated with medicines.
Brand:  Sharpsguard™ DD476OLR
 View more versions of this product
Product Code. 17754854
  • 22.15 € / Each
Edge
Explore more special offers
Description
Description
  • Sharpsguard™ eco 11.5 is a multi-purpose container featuring a robust pail handle for easy transportation. A wider aperture allows easy insertion of larger items.
  • This container is supplied with the base and lid separately. Details about how to lock the container and keep the users and contents safe can be found on the label.
  • It is designed to fit safely and securely into our POUDS™ mobile holders and bracket range.
  • This container is for disposal by incineration or other authorized process and is designed for the consignment of waste categorized by EWC codes: 18-01-01, 18-01-03, 18-02-01, 18-02-0
Specifications
Specifications
9.27 L
Orange
1 Ea.
275 mm
249 mm
622 g
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Sharpsguard™ Eco 11.5 Waste Container > Waste Container; Color: Orange