Learn More
Sharpsguard™ Eco 22 Waste Container
GREENER_CHOICE
The new Sharpsguard™ eco 22 container, made using at least 40% recycled plastic that is used for the disposal of sharps that have not been contaminated with medicines.
Brand:  Sharpsguard™ DD475OLR
 View more versions of this product
Product Code. 17734854
  • 31.30 € / Each
Edge
Explore more special offers
Description
Description
  • Sharpsguard™ eco 22 is ideal for theatres, laboratories and sluice rooms with high volumes of hazardous waste. The wide aperture allows economical and convenient disposal of large items.
  • This container is supplied with the base and lid separately. Details about how to lock the container and keep the users and contents safe can be found on the label.
  • It is designed to fit safely and securely into our POUDS™ mobile holders and bracket range.
  • This container is for disposal by incineration or other authorized process and is designed for the consignment of waste categorized by EWC codes: 18-01-01, 18-01-03, 18-02-01, 18-02-02.
Specifications
Specifications
17.86 L
Orange
1 Ea.
327 mm
343 mm
943 g