Learn More
Sharpsguard™ Eco 5 Waste Container
GREENER_CHOICE
The new Sharpsguard™ eco 5 containers, made using at least 40% recycled plastic that is used for the disposal of sharps that have not been contaminated with medicines.
Brand:  Sharpsguard™ DD471OLR
 View more versions of this product
Product Code. 17744844
  • 17.60 € / Each
Edge
Explore more special offers
Description
Description
  • Sharpsguard™ eco 5 is an ideal general-purpose sharps container, it provides maximum capacity in the minimum working space.
  • This container is supplied with the base and lid separately. Details about how to lock the container and keep the users and contents safe can be found on the label.
  • It is designed to fit safely and securely on our POUDS™ bracket range.
  • This container is for disposal by incineration or other authorized process and is designed for the consignment of waste categorized by EWC codes: 18-01-01, 18-01-03, 18-01-09, 18-02-01, 18-02-02, 18-02-08.
Specifications
Specifications
3.98 L
Orange
1 Ea.
176 mm
285 mm
338 g
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Sharpsguard™ Eco 5 Waste Container > Waste Container; Color: Orange