Learn More
Sharpsguard™ Eco Anatomical Waste Container
GREENER_CHOICE
The new Sharpsguard™ eco anatomical container, made using at least 40% recycled plastic with red solid lid is for the disposal of anatomical waste and other potentially infectious waste.
Brand:  Sharpsguard™ DD912R
 View more versions of this product
Product Code. 17764874
  • 154.00 € / Pack of 48
Explore more special offers
Description
Description
  • Sharpsguard™ eco anatomical is for the safe disposal of anatomical and other potentially infectious waste.
Specifications
Specifications
2.6 L
178 mm
166 mm
Red
Yellow
Safe Disposal of Anatomical and Potentially Infectious Waste