missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ Eco Best Technology™ Disposable Nitrile Gloves

Product Code. 17861181
Click to view available options
Size:
X-Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Eco Best Technology™ (EBT) biodegradable nitrile disposable gloves provide comfort, dexterity and performance across a wide range of industrial applications.

  • Same performance as non-EBT nitrile disposable gloves
  • Eco Best Technology™ (EBT) achieved 82% biodegradation in 386 days
  • Decreased risk of allergies
  • Second-skin fit and feel
  • Recommended for industrial applications including food, pharmaceutical, utilities, construction, agricultural and automotive

Compliance

Data Testing is available upon request.

Specifications

Certifications/Compliance ASTM D5511-18, ASTM D5526
Certifications/Compliance (Europe) EN 374-2:2003, EN 374-1:2016/Type B, EN 374-5:2016
Applications Industrial
AQL 1.5
Color Black
Disposable Single-use
Exterior Finish No Texture
Latex Free Yes
Material Nitrile
Size Large
Thickness (English) 4 mil
Thickness (Metric) 0.10 mm
Model 6112PFL
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'