missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ecolite(+) LC Fluid, MP Biomedicals
Liquid Scintillation Cocktails; Ecolite(+); MP Biomedical; Biodegradable
346.00 € - 786.00 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15742252
|
MP Biomedicals
0188247501 |
1 gal. |
346.00 €
3.79 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15752252
|
MP Biomedicals
0188247504 |
4 x 1 gal. |
786.00 €
15.14 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specifications
Liquid Scintillation Cocktails |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- ecolite
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P321-Specific treatment (see on this label).
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : phenylxylylethane, nonylphenol