Learn More
Cole Parmer™ Economical Polypropylene Graduated Cylinder
Polypropylene (PP) cylinders offer very good chemical resistance.
Brand:  Cole Parmer™ 06137-72
 View more versions of this product
Product Code. 15787968
  • 17.40 € / Each
Estimated Shipment: 24-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Meet ISO Standard 6706.
  • Calibrated to contain for measuring final volumes.
  • Molded graduations for durability.
  • Polypropylene construction provides very good chemical resistance.
Specifications
Specifications
Polypropylene
ISO Standard 6706