Learn More
Eutech™ EcoTestr pH Meter
Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications.
83.92 € - 97.28 €
Specifications
Accuracy (pH) | ±0.1 |
---|---|
Measurement Modes | pH |
Resolution (pH) | 0.1 |
IP Rating | IP67 |
Battery Type | A76/LR44 Alkaline |
Product Code | Brand | Calibration Points | For Use With (Application) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Calibration Points | For Use With (Application) | Price | Quantity & Availability | |||||
10462215
|
Eutech™
01X460901 |
1 | For pH 1 |
83.92 €
Each |
In Stock
Log in to see stock. ![]() |
|||||
11599054
|
Eutech™
01X460902 |
3 | For pH 2 |
97.28 €
Each |
In Stock
Log in to see stock. ![]() |
|||||
Description
From the market leader of testers comes the EcoTestr pH 1 & pH 2 – a new range of value-for-money, quality pH tester series designed for fuss-free pH measurements. Accurate, and easy to use, these testers are ideal for quick pH checks in hydroponic gardening, aquaculture, agriculture, pools, simple lab work and other water/wasterwater applications.
Features:
- 0.0 to 14.0 full pH range
- Auto-calibration & auto-buffer recognition
- IP67 waterproof – tester floats on water for easy retrieval
- Long sensor life – no need to change sensors
- Click-lock battery compartment requires no tools for battery replacements
- Hold function freezes reading temporarily for ease of recording
- Sensor protective cap that doubles up as a container for samples or calibration solution
Applications
Aquariums & aquaculture, hydroponics, swimming pools, spas & hot tubs, lab work and education, wastewater treatment, environmentmonitoring
Specifications
- pH Range: 0.0 to 14.0 pH
- Resolution: 0.1 pH
- Accuracy: ±0.1 pH
- Operating Temperature: 0 to 50°C
- Non-Volatilememory: Yes
- Hold Function: Yes
- Auto-Off: 8.5mins after last key pressed
- LCD Display: 3 ½ digit with annunciator
- Power: 4 X 1.5 V 'A76' micro alkaline batteries
- Ingress Rating: IP67 Waterproof
- Dimensions; Weight: Boxed: 24.5 cm x 13.5 cm x 4.5 cm; 137 g
Specifications
±0.1 | |
0.1 | |
A76/LR44 Alkaline |
pH | |
IP67 | |
250 hr. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.