missing translation for 'onlineSavingsMsg'
Learn More

Melford EDTA 0.5M Solution pH 8.0

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

Brand:  Melford E14000-250.0

Product Code. 30018248

  • 58.15 € / 250mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

  • Soluble in: Water
  • Store at: 2-8°C
Specifications

Specifications

EDTA 0.5 M Solution pH 8.0
6381-92-6, 1310-73-2, 7732-18-5
250 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers