missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDTA disodium salt dihydrate, BAKER ANALYZED™, J.T. Baker™
Click to view available options
Quantity:
250 g
500 g
Specifications
Specifications
Melting Point | 255°C |
pH | 4.0 - 6.0 (5% Solution at 25°C) |
Assay Percent Range | 99.0 to 101.0% |
Packaging | Plastic bottle for solids |
Quantity | 250 g |
Insoluble Matter | max. 0.005% |
Safety and Handling
Product Identifier
- EDTA disodium salt dihydrate BAKER ANALYZED Reagent
- Warning
- Acute toxicity Category 4
- Specific target organ toxicity after repeated exposure Category 2
- H332-Harmful if inhaled.
- H373-May cause damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P271-Use only outdoors or in a well-ventilated area.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P314-Get medical advice/attention if you feel unwell.
- MIXTURE LIST-Contains : EDTA disodium salt dihydrate
missing translation for 'provideContentCorrection'
missing translation for 'type03FormHeader'
missing translation for 'productTitle'