missing translation for 'onlineSavingsMsg'
Learn More

EDTA disodium salt solution, Volumetric, 0.05 M EDTA-Na2, Honeywell Fluka™

Product Code. 15603530
Click to view available options
Quantity:
500 mL
2.5 L

Volumetric, 0.05 M EDTA-Na2

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139-33-3
Molecular Formula C10H14N2Na2O8
Molecular Weight (g/mol) 336.21
MDL Number MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037
InChI Key ZGTMUACCHSMWAC-UHFFFAOYSA-L
Synonym ethylenediaminetetraacetic acid disodium salt, edta na2, disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid
PubChem CID 57339238
ChEBI CHEBI:64734
SMILES [Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

Quantity 500 mL
Linear Formula [-CH2N(CH2CO2Na)CH2CO2H]2
UN Number NONH for all modes of transport
Beilstein 3822669
IUPAC Name disodium;2-[2-[bis(carboxylatomethyl)azaniumyl]ethyl-(carboxylatomethyl)azaniumyl]acetate
Formula Weight 336.21g/mol
Density 1.01g/mL at 25°C
Chemical Name or Material Ethylenediaminetetraacetic Acid Disodium Salt Solution
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.