missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDTA Tetrasodium Salt Dihydrate, 99-101%, Ultrapure, Thermo Scientific Chemicals
71.70 € - 1256.00 €
Chemical Identifiers
CAS | 10378-23-1 |
---|---|
Molecular Formula | C10H20N2Na4O10 |
Molecular Weight (g/mol) | 420.233 |
MDL Number | MFCD00150026 |
InChI Key | DGWOWDJDIBPYOR-UHFFFAOYSA-N |
Synonym | ethylenediaminetetraacetic acid tetrasodium salt dihydrate, tetrasodium ethylenediaminetetraacetate dihydrate, edta-na4, edta tetrasodium salt, edta tetrasodium salt hydrate, ethylenediaminetetraacetic acid tetrasodium salt hydrate, 64-02-8 anhydrous, ksc909a7h, edta tetrasodium salt dihydrate, tetrapotassium ion 4-edta dihydrate |
PubChem CID | 131843362 |
IUPAC Name | 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid;sodium;dihydrate |
SMILES | C(CN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O.O.O.[Na].[Na].[Na].[Na] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15895278
|
Thermo Scientific
J15700.A1 |
1 kg | Plastic bottle |
71.70 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15805288
|
Thermo Scientific
J15700.C4 |
25 kg | Plastic bottle |
1256.00 €
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
10378-23-1 | |
420.233 | |
DGWOWDJDIBPYOR-UHFFFAOYSA-N | |
131843362 | |
C(CN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O.O.O.[Na].[Na].[Na].[Na] |
C10H20N2Na4O10 | |
MFCD00150026 | |
ethylenediaminetetraacetic acid tetrasodium salt dihydrate, tetrasodium ethylenediaminetetraacetate dihydrate, edta-na4, edta tetrasodium salt, edta tetrasodium salt hydrate, ethylenediaminetetraacetic acid tetrasodium salt hydrate, 64-02-8 anhydrous, ksc909a7h, edta tetrasodium salt dihydrate, tetrapotassium ion 4-edta dihydrate | |
2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid;sodium;dihydrate |
Specifications
10378-23-1 | |
C10H20N2Na4O10 | |
3861753 | |
DGWOWDJDIBPYOR-UHFFFAOYSA-N | |
2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid;sodium;dihydrate | |
131843362 | |
99 to 101% |
Crystalline Powder | |
MFCD00150026 | |
ethylenediaminetetraacetic acid tetrasodium salt dihydrate, tetrasodium ethylenediaminetetraacetate dihydrate, edta-na4, edta tetrasodium salt, edta tetrasodium salt hydrate, ethylenediaminetetraacetic acid tetrasodium salt hydrate, 64-02-8 anhydrous, ksc909a7h, edta tetrasodium salt dihydrate, tetrapotassium ion 4-edta dihydrate | |
C(CN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O.O.O.[Na].[Na].[Na].[Na] | |
420.233 | |
416.2 | |
Ethylenediaminetetraacetic Acid Tetrasodium Salt Dihydrate |
Safety and Handling
P201-P202-P261-P264b-P270-P271-P281-P301+P312-P304+P340-P305+P351+P338-P308+P313-P310-P330-P501c
H302+H332-H318-H350
missing translation for 'rtecsNumber' : AH4410000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only